Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-4-pyrimidinecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. H13921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13921G 1 g
H1392200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13921G Supplier TCI America Supplier No. H13921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6299-87-2
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00023267
InChI Key QYGHXDAYBIFGKI-UHFFFAOYSA-N
Synonym 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid
PubChem CID 239692
IUPAC Name 6-oxo-3,6-dihydropyrimidine-4-carboxylic acid
SMILES OC(=O)C1=CC(=O)N=CN1

Specifications

Melting Point 267°C
Quantity 1 g
Formula Weight 140.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-4-pyrimidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.