Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ 6-Hydroxy-DL-DOPA
Click to view available options
Quantity:
10 mg
Description
6-Hydroxy-DL-DOPA is an allosteric inhibitor of RAD52; inhibits RAD52 binding to single strand DNA binding domains (IC50 = 1.1 μM). Selectively inhibits proliferation of BRCA-deficient cancer cells in vitro. Also inhibits APE1.
Specifications
Specifications
Chemical Name or Material | 2,5-Dihydroxy-DL-tyrosine |
CAS | 21373-30-8 |
Quantity | 10 mg |
Target | DNA, RNA and Protein Synthesis Inhibitors |
Molecular Formula | C9H11NO5 |
Formula Weight | 213.19 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction