Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Methylthio)pyridine-3-boronic acid, 95%, Thermo Scientific™

Catalog No. aah27873md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27873MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH27873MD Supplier Thermo Scientific Chemicals Supplier No. H27873MD
Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321438-86-2
Molecular Formula C6H8BNO2S
Molecular Weight (g/mol) 169.005
MDL Number MFCD03788238
InChI Key UXWKVPJOPVIIRU-UHFFFAOYSA-N
Synonym 2-methylthiopyridine-5-boronic acid, 6-methylthio pyridin-3-ylboronic acid, 2-methylthio-5-pyridinyl-boronic acid, 2-methylthio-pyridine-5-boronic acid, 6-methylthio pyridin-3-yl boronic acid, 6-methylsulphanyl pyridine-3-boronic acid, 6-methylthio pyridin-3-yl-3-boronic acid, 6-methylthio pyridine-3-boronic acid, 5-borono-2-methylmercapto pyridine, 6-methylsulfanyl pyridin-3-ylboronic acid
PubChem CID 2762705
IUPAC Name (6-methylsulfanylpyridin-3-yl)boronic acid
SMILES B(C1=CN=C(C=C1)SC)(O)O

Specifications

Quantity 250 mg
Formula Weight 169.01
Percent Purity 95%
Chemical Name or Material 6-(Methylthio)pyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.