Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Nitropiperonyl alcohol, 98+%
Click to view available options
Quantity:
5 g
25 g
Description
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.
Notes
Light sensitive. Incompatible with oxidizing agents.
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 121°C to 125°C |
| Quantity | 5 g |
| Beilstein | 13749 |
| Sensitivity | Light sensitive |
| Formula Weight | 197.15 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 6-Nitropiperonyl alcohol |
Safety and Handling
| EINECSNumber | 239-376-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction