Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
60S Ribosomal Protein L26 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $437.74
Specifications
Antigen | 60S Ribosomal Protein L26 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE1438-25UL
|
MilliporeSigma
ABE143825UL |
25 μL |
Each for $192.35
|
|
|||||
ABE1438MI
|
MilliporeSigma
ABE1438 |
200 μL |
Each for $437.74
|
|
|||||
Description
Anti-60S Ribosomal Protein L26 Antibody is a highly specific rabbit polyclonal antibody that targets 60S ribosomal protein L26 and has been tested in Immunohistochemistry (Paraffin), and Western Blotting.Specifications
60S Ribosomal Protein L26 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
GST-tagged recombinant fragment corresponding to the first 95 amino acids from human 60S ribosomal protein L26. | |
Affinity Purified | |
This polyclonal antibody detects 60S ribosomal protein L26 in human. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Epigenetics and Nuclear Function | |
RPL26 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title