Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
7,7,8,8-Tetracyanoquinodimethane 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Quantity | 25 g |
UN Number | 3439 |
Formula Weight | 204.19 |
Percent Purity | ≥98.0% (HPLC,N) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 7,7,8,8-Tetracyanoquinodimethane |
Safety and Handling
EINECSNumber | (3)-1798 |
RTECSNumber | GU4850000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction