Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine, 97%, Thermo Scientific™

Catalog No. p-4509917
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 34950-82-8
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD09025906
InChI Key CYISPVTTZWJFEO-UHFFFAOYSA-N
Synonym 7-bromo-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine, 7-bromo-3,4-dihydro-2h-pyrido 3,2-b-1,4-oxazine, 7-bromo-2h,3h,4h-pyrido 3,2-b 1,4 oxazine, 7-bromo-2h,3h,4h-pyridino 2,3-e 1,4-oxazine, pubchem18740, acmc-1aimz, abbypharma ap-13-10238, 2h-pyrido 3,2-b-1,4-oxazine,7-bromo-3,4-dihydro
PubChem CID 24229635
SMILES BrC1=CN=C2NCCOC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 7-bromo-2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.