Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloro-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Catalog No. aah63693md
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82771-60-6
Molecular Formula C9H10ClN
Molecular Weight (g/mol) 167.64
MDL Number MFCD05861543
InChI Key BLRKQLICTKRDDZ-UHFFFAOYSA-N
Synonym 7-chloro-1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 7-chloro-1,2,3,4-tetrahydro, 7-chhloro-1,2,3,4-tetrahydroisoquinoline, 7-chloranyl-1,2,3,4-tetrahydroisoquinoline, isoquinoline,7-chloro-1,2,3,4-tetrahydro
PubChem CID 12595069
IUPAC Name 7-chloro-1,2,3,4-tetrahydroisoquinoline
SMILES ClC1=CC2=C(CCNC2)C=C1

Specifications

Quantity 250 mg
Formula Weight 167.64
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydroisoquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.