Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%

Catalog No. AAH6345606
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 73075-45-3 | C9H11Cl2N | 204.094 g/mol

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73075-45-3
Molecular Formula C9H11Cl2N
Molecular Weight (g/mol) 204.094
MDL Number MFCD08461076
InChI Key OGIAIXMUSSACDB-UHFFFAOYSA-N
Synonym 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 7-chloro-1,2,3,4-tetrahydro-, hydrochloride, 7-chloro-1,2,3,4-tetrahydro-isoquinoline hcl, 7-chhloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochlor, 7-chloro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 12595070
IUPAC Name 7-chloro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)Cl.Cl

Specifications

Quantity 5 g
Formula Weight 204.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.