Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

Catalog No. AC450460010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450460010 1 g
AC450460050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450460010 Supplier Thermo Scientific Chemicals Supplier No. 450460010

CAS: 799274-06-9 | C9H10FN·ClH | 187.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 799274-06-9
Molecular Formula C9H10FN·ClH
Molecular Weight (g/mol) 187.64
InChI Key LOJNQXIJLCPQDR-UHFFFAOYSA-N
Synonym 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-fluoro-1,2,3,4-tetrahydroisoquinoline hcl, 7-fluoro-1,2,3,4-tetrahydroisoquinolinehydrochloride, 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride salt, isoquinoline, 7-fluoro-1,2,3,4-tetrahydro-, hydrochloride, 7-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride salt, 7-fluoro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 45073971
IUPAC Name 7-fluoro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)F.Cl

Specifications

Melting Point 177.0°C to 179.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 187.64
Percent Purity 97%
Chemical Name or Material 7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.