Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Ketocholesterol Antibody (3F7), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP259357

 View more versions of this product

Catalog No. NBP259357


Only null left
Add to Cart

Description

Description

7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

7-Ketocholesterol
Monoclonal
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Mouse
Protein G purified
RUO
Specific for 7-ketocholesterol modified proteins.
Store at -20C.
IgG3
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
3F7
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
7KC, 7-KC, 7KC-Modified KLH
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μg
Primary
Non-species specific
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only