Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
7-Ketocholesterol Antibody (3F7), Novus Biologicals™

Mouse Monoclonal Antibody
$610.00
Specifications
Antigen | 7-Ketocholesterol |
---|---|
Clone | 3F7 |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
7-Ketocholesterol | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH). | |
Primary | |
Specific for 7-ketocholesterol modified proteins. |
3F7 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
7KC, 7-KC, 7KC-Modified KLH | |
IgG3 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title