Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Ketocholesterol Antibody (7E1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB259356700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259356700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259356700 Supplier Novus Biologicals Supplier No. NBP259356AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 7-Ketocholesterol
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 70
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 7KC, 7-KC, 7KC-Modified KLH
Host Species Mouse
Immunogen Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Specific for 7-ketocholesterol modified proteins.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.