Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxyindole, 97%, Thermo Scientific™

Catalog No. ac397520010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 3189-22-8
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00047203
InChI Key FSOPPXYMWZOKRM-UHFFFAOYSA-N
Synonym 7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi
PubChem CID 76660
IUPAC Name 7-methoxy-1H-indole
SMILES COC1=CC=CC2=C1NC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.1260g/mL
Boiling Point 166°C (27.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.126
Formula Weight 147.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 7-Methoxyindole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-690-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.