Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 719B Chemical Resistant Unsupported Nitrile Gloves

Catalog No. 17100808 Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Large
2X-Large

Chemical-resistant unsupported nitrile, 13 in., 11-mil, blue, raised diamond palm.

  • Protecting against a broad range of hazards, the SHOWA 719B chemical-resistant gloves features an extended gauntlet for forearm protection.
  • The 0.28 mm thick unsupported nitrile provides resistance against chemicals, solvents, oils, and animal fats.
  • The ergonomic design ensures comfort during long periods of wear.
  • Featuring a Diamond grip surface: makes wet work easier and safer.
  • 100% Nitrile
  • Unsupported
  • Unlined
  • Chemical Resistant
  • Water Resistant
  • Abrasion Resistant
  • Disposable
  • ForarmProtection
  • Oil Resistant

Specifications

Applications Laboratory
Color Blue
Finger Thickness (English) 11 mil
Finger Thickness (Metric) 0.025 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 11 mil
Palm Thickness (Metric) 0.025 mm
Product Type Chemical-resistant Gloves
Size Large
Thickness (English) 11 mil
Thickness (Metric) 0.025 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.