Learn More
Scott Safety™ 742 Series Air-purifying Respirator Accessory, Cartridges
Easy-breathingtwin cartridges were developed to provide optimal respiratory protection in non-IDLH industrial or HAZMAT situations
Products 9
Catalog Number Mfr. No. Cartridge Type Color FilterType Price Quantity & Availability  
Catalog Number Mfr. No. Cartridge Type Color FilterType Price Quantity & Availability  
19-150-1254
View Documents
Scott Safety™
7422SD1
Multipurpose/P100 Olive/Magenta Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-161-740
View Documents
Scott Safety™
7422SC1
Multipurpose Olive/Olive Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-161-796
View Documents
Scott Safety™
7422FP1
Particulate, P100 Cartridge Magenta/Magenta Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-161-797
View Documents
Scott Safety™
7422WA1
Acid Gas, P100 White/White Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-161-801
View Documents
Scott Safety™
7422BB1
Organic Vapor, P100 Black/Magenta Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-161-802
View Documents
Scott Safety™
7422YD1
Organic Vapor, Acid Gas, P100 Magenta/Yellow Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-161-800
View Documents
Scott Safety™
7422BA1
Organic Vapor Cartridge Black/Black Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-700-845
View Documents
Scott Safety™
7422WB1
Acid Gas, P100 Magenta/White Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-818-556
View Documents
Scott Safety™
7422GA1
Ammonia, Methylamine Green/Green Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
  • Low profile, swept back cartridge design ensures optimal field of vision and allows use of communications
  • Available in a full range of NIOSH approved, chemical cartridges or combination cartridges
  • Attaches directly to XCEL Halfmask and Promask full facepiece
  • Uses Quarter turn adapter when used with AV Series Facepieces
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.