Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl methanesulfonate, 99%
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.1600g/mL |
Boiling Point | 213.0°C to 214.0°C |
Flash Point | 100°C |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Refractive Index | 1.4170 to 1.4190 |
Linear Formula | CH3SO3C2H5 |
Quantity | 100 g |
Beilstein | 04,5 |
Specific Gravity | 1.16 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: May cause respiratory irritation. Causes serious eye irritation. May cause genetic defects. May cause cancer. Causes skin irritation. Harmful if swallowed. |
Precautionary Statement | GHS P Statement: Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 200-536-7 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction