Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ 8-Channel Micropipette
SDPSCI_ED

Catalog No. s39240 Shop All United Scientific Supplies, Inc Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
2 to 20 μL
20 to 200 μL
40 to 300 μL
5 to 50 μL

Catalog No. S39240

Supplier: United Scientific™ PMC8300

Only null left
Add to Cart
Add to Cart

The 8-Channel Micropipette is fully autoclavable and recommended for a wide variety of applications such as ELISA, molecular screening, kinetic studies, and DNA amplifications.

  • Multichannel micropipette.
  • Two-stop plunger.
  • Built-in tip ejector.
  • Autoclavable tip cone.
  • Click-stop adjustable digital display.
  • Easy to calibrate, maintain, and disassemble for autoclaving.
  • Manufactured in an ISO 9001:2008 certified facility.
  • Each unit calibrated according to ISO 8655 standards and includes a calibration certificate.

Specifications

Accuracy ±2.0% at 40.0 μL, ±1.0% at 150.0 μL, ±0.7% at 300.0 μL
Compatible Tips 1000 μL
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color White/Blue
Disposable Reusable
For Use With Micro Tip 1000 μL
Grip Type Ergonomic
Material Plastic
Model PMC8-300
Number of Channels 8
Programmability No
Sterility Non-sterile
Type Micropipette
Volume (Metric) 40 to 300 μL
For Use With (Application) Electrophoresis, PCR
For Use With (Equipment) Micropipette Tips
Includes Eight tips, One pipette holder
Precision ≤0.8% at 40.0 μL, ≤0.5% at 150.0 μL, ≤0.25% at 300.0 μL
Warranty 1 Year
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.