Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Methoxypsoralen, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
298-81-7
C12H8O4
216.19
MFCD00005009
QXKHYNVANLEOEG-UHFFFAOYSA-N
methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
4114
CHEBI:18358
9-methoxyfuro[3,2-g]chromen-7-one
COC1=C2OC=CC2=CC2=C1OC(=O)C=C2
298-81-7
C12H8O4
216.19
MFCD00005009
QXKHYNVANLEOEG-UHFFFAOYSA-N
methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
4114
CHEBI:18358
9-methoxyfuro[3,2-g]chromen-7-one
COC1=C2OC=CC2=CC2=C1OC(=O)C=C2
Water 0.5% max. (K.F.)
Packaging Glass bottle
Quantity 5g
Melting Point 143.0°C to 148.0°C
Infrared Spectrum Authentic
Color White to Cream
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (on anhydrous substance) (HPLC)
Beilstein 19, I, 711
Merck Index 15, 6059
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: sparingly soluble in boiling water,sparingly soluble in ether,soluble in boiling alcohol,acetone,acetic acid,,propylene glycol and benzene,freely soluble in chloroform
Formula Weight 216.19
Physical Form Needle-Like Crystals or Powder
Percent Purity 99%
Chemical Name or Material 8-Methoxypsoralen, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-066-9

RTECSNumber : LV1400000

TSCA : TSCA