Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Methoxypsoralen, 99%, MP Biomedicals™

Catalog No. ICN10329601
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 298-81-7
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00005009
InChI Key QXKHYNVANLEOEG-UHFFFAOYSA-N
Synonym methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
PubChem CID 4114
ChEBI CHEBI:18358
IUPAC Name 9-methoxy-7H-furo[3,2-g]chromen-7-one
SMILES COC1=C2OC=CC2=CC2=C1OC(=O)C=C2

Specifications

Color White to Cream
Assay Percent Range 0.99
Quantity 1 g
Formula Weight 216.2
Physical Form Powder
Chemical Name or Material 8-Methoxypsoralen
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.