Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 8-pCPT-2-O-Me-cAMP-AM
GSA_VAGREENER_CHOICESDP

Catalog No. 4853100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-531-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-531-00U Supplier Tocris Bioscience™ Supplier No. 4853/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Epac activator; cell-permeable analog of 8CPT-2Me-cAMP (Cat. No. 1645)

Selective Epac activator; cAMP analog. Induces Rap activation and junction tightening in HUVECs; triggers adhesion of Jurkat-Epac1 cells to fibronectin. Stimulates insulin secretion in rat INS-1 cells. More potent, cell-permeable analog of 8CPT-2Me-cAMP (Cat. No. 1645).

This product is a mixture of axial and equatorial isomers. Both isomers give 8CPT-2Me-cAMP after esterase cleavage.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1152197-23-3
Molecular Formula C20H21ClN5O8PS

Specifications

Quantity 100 μg
Formula Weight 557.9
Purity >97%
Molecular Weight (g/mol) 557.899
Chemical Name or Material EPAC Activators
Recommended Storage Store at -80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.