Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Quinolinol, Crystal, ACS, Spectrum™ Chemical
SDP

$196.00 - $6990.00

Specifications

Grade ACS
CAS 148-24-3
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-608-299
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H1095125GM
125 g Amber Glass Bottle
Each for $323.00
Only null left
Add to Cart
 
18-608-300
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H109512KG
12 kg Poly Pail
Each for $3,163.00
Only null left
Add to Cart
 
18-608-301
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H1095500GM
500 g Amber Glass Bottle
Each for $663.00
Only null left
Add to Cart
 
18-608-302
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H109550KGBL
50 kg Fiber Drum
Each for $6,990.00
Only null left
Add to Cart
 
18-608-303
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H10952.5KG
2.5 kg Poly Pail
Each for $2,091.00
Only null left
Add to Cart
 
18-608-304
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H109525KGBL
25 kg Fiber Drum
Each for $4,224.00
Only null left
Add to Cart
 
18-608-305
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H109525GM
25 g Amber Glass Bottle
Each for $196.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

ACS
148-24-3
Videos
SDS
Documents

Documents

Product Certifications

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.