Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek 800 TS Absorbance Reader PROMO

Catalog No. B800TSUVW25
Click to view available options
Model:
800TS
800TSI
800TSNB
800TSUV
800TSUVI
This item is not returnable. View return policy
This item is not returnable. View return policy

Affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays

The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Microplate Reader
Detector Type Photodiode
Interface USB Type B, USB Type A
Light Source Tungsten Halogen Lamp
Reading Options Endpoint, Kinetic, Well-area Scanning
Shaking Linear
Software Type Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Bandwidth 10 nm
Certifications/Compliance CE and TUV marked. RoHS Compliant.
Filters 340, 405, 450, 490, 630 nm
For Use With (Application) Measuring Absorbance in Microplate-based Assays
Includes 6- to 96-well plates, 340 to 750 nm, Shaking
Model 800TSUV
Voltage 100/240 V
Wavelength Range 340 to 750 nm
Weight (English) 18.5 lb.
Weight (Metric) 8 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.