Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Showa™ 8005PF Powder-Free Nitrile Glove

Showa™ Powder-Free Nitrile Glove is a powder-free option that is ideal for jobs requiring the utmost in barrier protection, while keeping tactile discrimination.

Specifications

Certifications/Compliance FDA Food Compliance Approved
Certifications/Compliance (Europe) CE Cat. III 2777, EN ISO 374-1:2016/Type B (JKO), EN ISO 374-5:2016
Color Blue
Hand Ambidextrous
Material Nitrile
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-020-285
View Documents
Showa™
8005PFSIND
Small Pack of 50 for $44.20
Case of 20 PK for $397.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-020-286
View Documents
Showa™
8005PFMIND
Medium Pack of 50 for $43.65
Case of 20 PK for $392.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-020-287
View Documents
Showa™
8005PFLIND
Large Pack of 50 for $44.55
Case of 20 PK for $401.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-020-288
View Documents
Showa™
8005PFXLIND
X-Large Pack of 50 for $44.40
Case of 20 PK for $399.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Easy to tear away in case of emergency break
  • Can easily be one-hand use
  • Rolled cuff prevents dirt from entering the glove
  • Second-skin feel significantly reduces hand fatigue
  • Latex-free
  • 100% nitrile
  • Powder free
  • Low-modulus formulation
  • Rolled cuff
  • Applications:
    • Biotechnology
    • Pet Food Production
    • Washing and cleaning
    • Coroner/morgue
    • Public utility
    • Life sciences
    • Pharmaceuticals and API
Specifications

Specifications

FDA Food Compliance Approved
Blue
Nitrile
0.20 mm
CE Cat. III 2777, EN ISO 374-1:2016/Type B (JKO), EN ISO 374-5:2016
Ambidextrous
8 mil
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit