Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ BRAND™ Deep Well Plates
Click to view available options
Volume (Metric) Well:
0.3 mL
0.5 mL
1.1 mL
1.2 mL
2.2 mL
Format:
Standard
Well Shape:
Round
Round Bottom (U-well)
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
Description
BRAND™ Deep Well Plates are designed for a broad range of applications, including sample storage, high-throughput screening (HTS), cell and tissue culture, serial dilutions, and reagent transfers. Most plates are manufactured from high-quality virgin polypropylene, with one highly transparent polystyrene variant for applications requiring optical clarity.
- Available in 96- and 384-well formats
- Manufactured from high-quality virgin polypropylene or polystyrene
- BIO-CERT™ PCR Quality are free from human DNA, DNase, RNase, and pyrogens
- Polypropylene plates are autoclavable, chemically resistant, and suitable for storage down to -80°C (-112°F)
- Polystyrene plate offers excellent transparency and is suitable for storage down to -20°C (-4°F)
- ANSI/SLAS 1 and 4 compliant for compatibility with automated systems
- Alphanumeric well labeling and stackable design for efficient organization
- Notched corners ensure correct orientation in robotic and automated liquid-handling systems
- Compatible with sealing mats or adhesive films to minimize evaporation and contamination

Specifications
Specifications
Barcode | No Barcode |
Format | Standard |
Array | 8 x 12 |
Color | Clear |
Product Type | 96-well Assay Microplate |
Material | Polypropylene |
Volume (Metric) Well | 1.2 mL |
No. of Wells | 96 |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction