Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
GREENER_CHOICE

Catalog No. e0030129610 Shop All Eppendorf North America Products
Click to view available options
Color:
Blue
Colorless
Green
Red
Yellow
Skirt Style:
Semi-skirted
Unskirted
Unskirted, Divisible
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
PCR Clean and Free of Bacterial DNA (Non-sterile)
Packaging:
5 Bags x 4 Plates
5 Bags x 5 Plates
Individually Blister Wrapped

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.

Certifications

Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Individually Blister Wrapped
Color Colorless
Product Type PCR Plate
Skirt Style Semi-skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 250 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) PCR with medium to high sample throughput
Max. RCF 2250 x g
Purity or Quality Grade Forensic DNA Grade (Non-sterile)
Quantity 10 Pk.
Rimmed Yes
Well Color Colorless
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.