Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Absolute Q™ MAP16 Plate Kit and Absolute Q™ Universal DNA dPCR Master Mix

Catalog No. A40001697
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40001697 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40001697 Supplier Applied Biosystems™ Supplier No. A40001697
Add to Cart
Add to Cart

Eliminate the need to design custom assays and choose from over 20 million predesigned assays. Includes the Absolute Q MAP16 plate kit plus the Absolute Q Universal DNA dPCR master mix which is compatible with a wide range of applications and difficult targets.

The Absolute Q MAP16 Plate Kit and Absolute Q Universal DNA Digital PCR Master Mix are key components of the QuantStudio Absolute Q Digital PCR System. The system is powered by microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV).

Absolute Q MAP16 Plate Kit

The Absolute Q MAP16 microinjection-molded plates overcome common challenges of inconsistency and high dead volume. This novel approach provides many benefits, including precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed.

Absolute Q Universal DNA Digital PCR Master Mix

This package product also includes Absolute Q Universal DNA Digital PCR Master Mix, which is compatible with a broad range of applications and over 20 million predesigned TaqMan Assays (performance-guaranteed*), reducing the need to design custom assays. Simplify your workflow by avoiding assay design and minimizing assay optimization; use the same assay for both qPCR and digital PCR. In addition, with 72 hours of assembled-reaction benchtop stability, this master mix enables increased productivity through automation.

The impressive precision and accuracy of the Absolute Q system helps provide confidence in measurements across replicates and experiments.

* We guarantee that our predesigned TaqMan Assays will enable you to obtain data you can trust, or we'll replace them at no cost or give you a credit for the assay purchase price. Terms and conditions apply.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 12 MAP16 Plates, Gaskets, Isolation Buffer, and 1 tube Absolute Q™ Universal DNA Digital PCR Master Mix (192 reactions)
Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance High
PCR Method Digital PCR
Reaction Speed Fast or Standard
Quantity 1 Ea.
For Use With (Application) Digital PCR (dPCR), DNA Quantitation, Gene Expression, Genotyping, Microbial Detection
No. of Reactions 192 Reactions
How many MAP16 plates are included in the Absolute Q MAP16 Plate Kit and Absolute Q Universal DNA dPCR Master Mix (Cat. No. A40001697)

12 packs of MAP16 plates.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.