Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloropyrimidine-5-carboxylate, 97%

Catalog No. AAH6491406
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6491406 5 g
AAH6491403 1 g
AAH64914MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6491406 Supplier Thermo Scientific Chemicals Supplier No. H6491406
Add to Cart

CAS: 89793-12-4 | C7H7ClN2O2 | 186.595 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89793-12-4
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.595
MDL Number MFCD09863164
InChI Key IEMKQRSOAOPKRJ-UHFFFAOYSA-N
Synonym 2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 2-chloro-5-ethoxycarbonyl pyrimidine, ethyl-2-chloropyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, ethyl2-chloropyrimidine-5-carboxylate, acmc-209r2w, ksc663i8r, ethyl 2-chloranylpyrimidine-5-carboxylate, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester
PubChem CID 10487815
IUPAC Name ethyl 2-chloropyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1)Cl

Specifications

Melting Point 52°C to 60°C
Boiling Point 80°C (760 mmHg)
Quantity 5 g
Beilstein 142651
Formula Weight 186.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 2-chloropyrimidine-5-carboxylate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.