Learn More
Agilent BioTek 50 TS Microplate Washer, IVD
GREENER_CHOICE
The BioTek 50 TS washer offers excellent microplate washing performance. For In Vitro Diagnostic Use.
Supplier:  Agilent BioTek 50TS12SI
 View more versions of this product
Catalog No. BT50TS12SI
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Description
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE and TUV marked. RoHS compliant.
Optional (2 to 4 L)
Programmable in minutes and seconds, up to 30 min.
9.8 kg
25 to 3000 μL/well
150 to 1000 μL/well/sec. (manifold dependent)
50TS12
4.3 in. Color LCD Touch Screen Display
USB (1)
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Microplate Washer
1 to 10
4.3 in. LCD Color Touch Screen
One positive displacement syringe drive
96-well plates only
22 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.