Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Racked Filtered Micropipette Tips
Shop All Eisco Products

Click to view available options
Volume (Metric):
0.2 to 10 μL
2 to 100 μL
2 to 20 μL
2 to 200 μL
2 to 50 μL
200 to 1000 μL
50 to 300 μL
Description
- Sterile
- Graduated
- Contain hydrophobic polyethylene filter
- Free of lubricants, dyes and heavy metals
- 96 tips supplied in reusable racks
- Rack and tips are autoclavable
- Made of plastic
- Free of lubricants, dyes, and heavy metals
Specifications
Specifications
Description | Pack of 96 |
Volume (Metric) | 50 to 300 μL |
Filtered | Filtered |
Sterility | Sterile |
Color | Clear |
Rack Color | Yellow |
Autoclavable | Autoclavable |
For Use With (Application) | Electrophoresis, PCR |
Format | Rack |
Tip Style | Fine Point |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction