Learn More
United Scientific™ Knife Edge Lever Clamp
Shop All United Scientific Supplies, Inc Products
Click to view available options
Color:
Silver
Description
- Can be attached to any standard meter stick.
- Used as a knife-edge fulcrum for supporting the meter stick in the inverted position.
- Prop 65 Warning Label.
Specifications
Specifications
| Type | Lever Clamp |
| Material | Steel |
| Color | Silver |
| Jaw Type | Single Adjustment |