Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Wide Mouth Media/Storage Bottles
SDPSCI_ED

Catalog No. S27789 Shop All United Scientific Supplies, Inc Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
5000 mL

Catalog No. S27789

Supplier: United Scientific Supplies BSWM5000

Only null left
Add to Cart
Add to Cart

Square-shaped media bottles made of borosilicate glass have large, wide-mouth openings that enable easy storage and transfer of substances

  • Borosilicate Glass Bottles
  • Wide mouth for easy transfer and storage of substances
  • Easy-to-read printed graduations
  • Includes GL80 polypropylene screw caps
  • Autoclavable

Specifications

Capacity (Metric) 5000 mL
Capacity (English) 169 oz.
Color Clear
Includes GL80 Polypropylene Screw Caps
Material Borosilicate Glass
Packaging Each
Quantity 1
Sterility Non-sterile
Closure Included Yes
Graduated Yes
Mouth Threaded
Shape Square
Product Type Storage Bottle
Closure Material Polypropylene
Closure Size GL 80
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.