Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Wide Mouth Glass Reagent Bottle with Glass Stopper
Shop All Eisco Products

Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
24/29 mm
29/32 mm
34/35 mm
45/40 mm
50/42 mm
55/44 mm
Description
Warranty and Services
1 year limited manufacturer warranty
Specifications
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 17 oz. |
Color | Clear |
Description | Wide neck |
For Use With (Application) | Holding liquids |
Includes | Interchangeable, tight fitting, hollow glass stopper (socket size 24/29) |
Material | Borosilicate Glass |
Length (Metric) | 10.2 cm |
Quantity | Pack of 1 |
Sterility | Non-sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction