Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT-VirusGEN™ Transfection Reagent
Shop All Mirus Bio LLC Products
Click to view available options
Quantity:
1 x 0.3 mL
1 x 0.75 mL
1 x 1.5 mL
5 x 1.5 mL
10 x 1.5 mL
Description
TransIT-VirusGEN™ Transfection Reagent is designed to enhance delivery of packaging and transfer vectors to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
- Reliable – Consistent high virus titer production
- Scalable – Efficient across different formats
- Flexible – Compatible with different recombinant virus and cell culture systems
- Animal Origin Free – High performance with maximum compatibility
Specifications
Specifications
Content And Storage | Store at -10°C to -30°C |
Description | Before each use, warm to room temperature and vortex gently. |
Cell Type | 293-derived Cells |
Format | Tube |
Promoter | None (Promoterless) |
Transfection Technique | Chemical Transfection |
Product Type | Transfection Reagent |
Quantity | 1 x 0.3 mL |
Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction