Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Benzene Ring Molecular Model Kit
SCI_ED

Catalog No. s139062 Shop All Walter Products Products
Change view
Click to view available options
Grade:
6+
Catalog No. Grade
S139062 6+
1 options

Catalog No. S139062

Supplier: Walter Products™ MOLE1014

Only null left
Add to Cart
Add to Cart

An excellent visual aid to learn the geometry and bond angles of benzene!

This durable and colorful plastic model of benzene is excellent for hands-on learning and visual display.

  • Made of durable and colorful plastic
  • Molecular geometry and bond angles can be visibly seen in the structure, making it great for studying
  • Great molecular model kit for students studying chemistry in middle school or university
  • A common group of organic compounds

Specifications

For Use With (Application) Organic Chemistry
Grade 6+
Material Plastic
Product Type Molecular Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.