Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Tris-Compatible Flat pH Sensor
SDPSCI_ED

Catalog No. s15956nd Shop All Vernier Software Products
Click to view available options
Temperature Range (Metric):
0°C to 100°C

Includes

Tris-Compatible Flat pH Electrode BNC, Electrode Amplifier, pH Storage Solution

Requires

Requires an interface and software, sold separately

Use the Tris-Compatible Flat pH Sensor to measure the pH of a solution or semisolid, such as food or soil.

Range: pH 0–14 (Some sodium error in ranges greater than pH 12 due to thicker glass and higher impedance values) Electrode type: Double-junction, sealed, gel-filled, Ag/AgCl reference, polycarbonate body Membrane style: Flat glass Storage solution: pH 4/KCl solution (10 g KCl in 100 mL buffer pH-4 solution) Cable: 1 meter coaxial cable with BNC connector Temperature range: 0– 100ºC 12 mm OD Impedance: ∼20 kΩ at 25ºC Response time: 98% of full response in 30 s at 25ºC

Warranty and Services

5-year limited warranty

Specifications

Grade High School and College
Warranty 5 Year Limited
For Use With (Application) Educational use only
Range 0 to 14 pH
Response Time 98% of full response in 30 s at 25°C
Temperature Range (Metric) 0°C to 100°C
Length (Metric) 1 m (Cable)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.