Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Gas Pressure Sensor
SDPSCI_ED

Catalog No. s16052nd Shop All Vernier Software Products
Click to view available options
Grade:
K-8, High School, College

Includes

Vernier Gas Pressure Sensor, Tapered valve connectors (2) inserted into a No. 5 stopper, Tapered valve connector (1) inserted into a No. 1 stopper, Two-way valve, 18 inch Tygon tubing with Luer Lock connectors, 20 mL syringe, White transpiration tubing clamps (2)

Requires

Requires an interface and software, sold separately

Use the Gas Pressure Sensor to monitor pressure changes of a gas in chemistry and biology experiments.

Pressure Range: 0 to 210 kPa (0 to 2.1 atm or 0 to 1600 mm Hg) Accuracy: ±4 kPa Maximum pressure that the sensor can tolerate without permanent damage: 4 atm Sensing Element: Honeywell SSCMRNN030PAAA5

Warranty and Services

5-year limited warranty

Specifications

Grade K-8, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy ±4 kPa
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.