Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Differential Voltage Probe
SDPSCI_ED

Catalog No. s16060nd Shop All Vernier Software Products
Click to view available options
Grade:
K-8, High School, College

Includes

Vernier Differential Voltage Probe

Requires

Requires an interface and software, sold separately

The Differential Voltage Probe is designed for exploring the basic principles of electricity. Use this probe to measure voltages in low voltage AC and DC circuits.

Input voltage range: ±6.0 V Maximum voltage on any input: ±10 V Input Impedance (to ground): 10 M Ω Differential Impedance: >20 M Ω Typical Resolution: 3.1 mV Linearity: 0.01% Supply voltage: 5 VDC

Warranty and Services

5-year limited warranty

Specifications

Grade K-8, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 3.1 mV
Electrical Requirements 5 VDC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.