Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Force Plate
SDPSCI_ED

Catalog No. s16152nd Shop All Vernier Software Products
Click to view available options
Grade:
Middle School, High School, College

Includes

Vernier Force Plate, (1) Pair of handles

Requires

Requires an interface and software, sold separately

Designed to collect data on forces developed during stepping, jumping, and other human-scale actions.

Force range: –850 to +3500 N or –200 to +850 N, where positive value is a compression force Maximum non-damaging force: 4500 N (1000 lb) compression or 900 N (200 lb) pull evenly distributed Typical Resolution: 1.2 N or 0.3 N Dimensions: 28 cm by 32 cm by 5 cm

Warranty and Services

5-year limited warranty

Specifications

Grade Middle School, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 1.2 N or 0.3 N
Dimensions (L x W x H) 28 x 32 x 5 cm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.