Learn More
Vernier™ Go!Link
SDPSCI_ED
Catalog No. s16161nd Shop All Vernier Software Products
Click to view available options
Warranty:
5 Year Limited
Catalog No. S16161ND
Supplier: Vernier™ GOLINK
Only null left
Add to Cart
Add to Cart
Includes
Vernier Go!Link interface, Quick-start guide, Data-Collection Software, Logger Lite and Graphical Analysis for Chrome are free downloads., Ordering Information, This product is discontinued and no longer available.
Vernier’s low-cost USB sensor interface connects over 40 Vernier sensors to a Windows or Macintosh computer.
  • USB specification: 1.1
  • Maximum sample rate: 200 samples per second
  • a Qquick and affordable way to get started with data-collection technology.
  • It's a single-channel interface that connects most Vernier sensors to your computer or Chromebook USB port.
Warranty and Services
5-year limited warranty
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 12 bit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.