Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vernier™ Light Sensor

Shop All Vernier Software Products


Click to view available options
Grade:
Middle School, High School, College
Description
Includes
Vernier Light Sensor
Requires
Requires an interface and software, sold separately
Resolution 0–600 lux: 0.2 lux 0–6,000 lux: 2 lux 0–150,000 lux: 50 lux Wavelengths: 400–800 nm
Warranty and Services
5-year limited warranty
Specifications
Specifications
| Description | Wavelength range 400 to 800 nm |
| Grade | Middle School, High School, College |
| Interface | Required |
| Warranty | 5 Year Limited |
| For Use With (Application) | Educational use only |
| Resolution | 0.2 lux at 0 to 600 lux, 2 lux at 0 to 6000 lux, 50 lux at 0 to 150000 lux |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction