Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Basalt Specimen
SCI_ED

Catalog No. S26643 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26643 1 Ea. 7 to 8 cm
S24748 1 Ea. 2 to 3 cm
S26589 10/Pk. 2 to 3 cm
S24749 12/Pk. 2 to 3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26643 Supplier Eisco™ Supplier No. ESNG0041HAND
Add to Cart
Add to Cart

Basalt is formed quickly from an iron-rich magma and exhibits a dark color and small crystal size.

  • Aphanitic volcanic rock forms from effusive eruptions, such as shield volcanoes of Hawaii, or deep ocean floor
  • For use in introduction to igneous rock classification and identification
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Students can recreate cooling history of this sample based on texture and abundance of phenocrysts
  • Example of aphanitic igneous rock

Specifications

Product Type Igneous Rock
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Specimen Type Basalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.