Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Clean-Trace™ Biomass Detection Kit ULW10, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. s27253 Shop All AquaPhoenix Scientific Products
Click to view available options
:
Biomass Detection Kit ULW10, Sold by AquaPhoenix Scientific

Water test kit using ATP bioluminescence to detect overall biological load in a sample

  • For testing aqueous samples requiring filtration and/or extraction such as viscous samples or ones that contain particulates or fibers
  • Assesses amount of biomass in process waters, waste waters, water tanks and cooling towers by determining total ATP present
  • Helps determine biological status of process equipment and effects of biocide treatments

Specifications

Format Kit
For Use With (Application) Detecting biological load in a sample
No. per Case 100/Cs.
Type Biomass detection kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.