Learn More
3M™ Clean-Trace™ Hygiene Monitoring and Management System LM1 ATP System, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. s30761 Shop All AquaPhoenix Scientific Products
Click to view available options
Type:
Instrument
Hand held diagnostic tool to verify cleaning effectiveness to capture, store and manage data for later analysis
  • Re-engineered for improved precision and accuracy
  • Easy to use and fast time to result
  • Robust for the manufacturing environment
  • System includes the 3M™ Clean-Trace™ Luminometer and 3M™ Clean-Trace™ Hygiene Monitoring and Management Software to help monitor testing results over time
  • Ergonomic design for simple one-handed operation
  • Wireless data transfer
  • 23 languages for multilingual pre-production teams
  • Robust to provide consistent results across temperatures
  • New software has easy-to-use dashboard that immediately turns data into actionable information
Specifications
Type Instrument
For Use With (Application) Monitoring hygiene and analyzing data
Includes 3M™ Clean-Trace™ Luminometer, 3M™ Clean-Trace™ Hygiene Monitoring and Management Software
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.