Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products Large Microscope Carrying Case
SCI_ED

Catalog No. s80995 Shop All Walter Products Products
Change view
Click to view available options
Material:
Nylon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S80995 Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80995 Supplier Walter Products Supplier No. CC100
Only null left
Add to Cart
Add to Cart

Thick, reinforced, nylon built case that has padded cushioning inside and velcro straps to secure your microscope in place

Fits microscopes with trinocular heads

Specifications

For Use With (Application) Education
Dimensions (L x W x H) 12.5 x 11 x 19 in.
Color Black
Includes Microscope Case
For Use With (Equipment) Microscope with trinocular heads
Holds 1 Microscope
Material Nylon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.