Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ A 196

Click to view available options
Quantity:
10 mg
50 mg
Description
A 196 is a potent and selective SUV420H1/H2 inhibitor (IC50 values are 25 and 144 nM respectively) that exhibits 100-fold selectivity over other histone methyltransferases and non-epigenetic targets. A 196 inhibits the di- and tri-methylation of H4K20me in multiple cell lines (IC50 < 1 μM).
To request the negative control for A 196, please fill out the SGC 2043 request form on the SGC website.
Specifications
Specifications
Chemical Name or Material | 6,7-Dichloro-N-cyclopentyl-4-(4-pyridinyl)-1-phthalazinamine |
CAS | 1982372-88-2 |
Assay Percent Range | >98% |
Quantity | 10 mg |
Molecular Formula | C18H16Cl2N4 |
Formula Weight | Observed MW: 359.25 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction