Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
A 77-01 is a potent TGF-βRI (ALK-5) inhibitor (IC50 = 34 nM). Inhibits TGF-β transcriptional activation. Inhibits TGF-β-induced proliferation of lung epithelial cells and SMAD phosphorylation. Inhibits TGF-β-induced epithelial-to-mesenchymal transition (EMT). Also inhibitor of p38 MAPK. Likely active metabolite of A 83-01 (Cat. No. 2939).
Greener Choice Claims
- Environmental benefits include:
- Ozone-Safe or Safe for the Ozone Layer
Specifications
Specifications
Chemical Name or Material | 4-[3-(6-Methyl-2-pyridinyl)-1H-pyrazol-4-yl]quinoline |
CAS | 607737-87-1 |
Quantity | 25 mg |
Molecular Formula | C18H14N4 |
Formula Weight | 286.33 |
Purity | 98% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience⢠A 77-01 >
Spot an opportunity for improvement?Share a Content Correction