Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 83-01
GSA_VAGREENER_CHOICE

Catalog No. p-4783704
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of TGF-βRI, ALK4 and ALK7

A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I activin/nodal receptor ALK4 and type I nodal receptor ALK7 (IC50 values are 12, 45 and 7.5 nM respectively). A 83-01 blocks phosphorylation of Smad2 and inhibits TGF-β-induced epithelial-to-mesenchymal transition. Only weakly inhibits ALK-1, -2, -3, -6 and MAPK activity. More potent than SB 431542 (Cat. No. 1614). A 83-01 inhibits differentiation of rat induced pluripotent stem cells (riPSCs) and increases clonal expansion efficiency. Helps maintain homogeneity and long-term in vitro self-renewal of human iPSCs. Also promotes neural differentiation of hPSCs as part of a chemical cocktail.

For more information about how A 83-01 may be used, see our protocol: Converting Fibroblasts into Cardiomyocytes (9C Cocktail)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 909910-43-6
Molecular Formula C25H19N5S

Specifications

Quantity 10 mg
Formula Weight 421.52
Purity >98%
Molecular Weight (g/mol) 421.522
Chemical Name or Material A 83-01
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.