Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 867744
GREENER_CHOICE

Catalog No. p-4816870
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of α7 nAChR

Positive allosteric modulator of α7 nAChRs (IC50 values are 0.98 and 1.12 μM for human and rat α7 receptor ACh-evoked currents respectively, in X. laevis oocytes). Displays no activity at 5-HT3A, α3β4 or α4β2 nAChRs. Brain penetrant.

Chemical Identifiers

CAS 1000279-69-5
Molecular Formula C20H19ClN2O3S

Specifications

Quantity 10 mg
Formula Weight 402.89
Purity >97%
Molecular Weight (g/mol) 402.893
Chemical Name or Material A 867744
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.