Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ A 967079
GREENER_CHOICE

Catalog No. 471610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-161-0 10 mg
47-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-161-0 Supplier Tocris Bioscience™ Supplier No. 4716/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TRPA1 channel blocker

Selective TRPA1 channel blocker (IC50 values are 67 and 289 nM at human and rat TRPA1 receptors respectively). Displays 1000-fold selectivity for TRPA1 over other TRP channels, and is >150-fold selective over 75 other ion channels, enzymes and G-protein-coupled receptors. Exhibits analgesic effects in a rat osteoarthritic pain model.

Chemical Identifiers

CAS 1170613-55-4
Molecular Formula C12H14FNO

Specifications

Quantity 10 mg
Formula Weight 207.24
Purity >98%
Molecular Weight (g/mol) 207.248
Chemical Name or Material A 967079
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.